Learn More
Sino Biological APRIL / cd256 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100641T40100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human APRIL/cd256, and purified by antigen affinity chromatography.
Catalog No. 50-161-1717