Learn More
Sino Biological Human ALK-1 / ACVRL1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10066H08H200
A DNA sequence encoding the the extracellular domain of human ALK1 (NP_000011.2) (Met 1-Gln 118) was fused with a polyhistide tag at the C-terminus.
Catalog No. 50-161-3930