Learn More
Sino Biological ALK-1 / ACVRL1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10066RP0250
Produced in rabbits immunized with purified, recombinant Human ALK-1 / ACVRL1 (rh ALK-1 / ACVRL1; Catalog#10066-H08H; NP_000011.2; Met1-Gln118). ALK-1 / ACVRL1 specific IgG was purified by Human ALK-1 / ACVRL1 affinity chromatography.
Catalog No. 50-161-1780