Learn More
Sino Biological COX4I1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100663T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human COX4I1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1747