Learn More
Sino Biological RAB9A / RAB9 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100671T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human RAB9A, and purified by antigen affinity chromatography.
Catalog No. 50-161-1784