Learn More
Sino Biological Pan Cytokeratin / KRT(pan) Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100675T36200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Pan Cytokeratin/KRT(pan), and purified by antigen affinity chromatography.
Catalog No. 50-161-1797