Learn More
Sino Biological Carboxypeptidase E / CPE Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10069R001100
This antibody was obtained from a rabbit immunized with purified, recombinant Human CPE (rhCPE; Catalog#10069-H08H; NP_001864.1; Met 1-Ser 453).
Catalog No. 50-161-1838