Learn More
Sino Biological Carboxypeptidase E / CPE Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10069RP01100
Produced in rabbits immunized with purified, recombinant Human CPE (rhCPE; Catalog#10069-H08H; NP_001864.1; Met 1-Ser 453). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-1840