Learn More
Sino Biological Carboxypeptidase E/CPE Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10069T40100
Produced in rabbits immunized with purified, recombinant Human Carboxypeptidase E/CPE (rh Carboxypeptidase E/CPE; Catalog#10069-H08H; NP_001864.1; Met1-Ser453). Carboxypeptidase E/CPE specific IgG was purified by Human Carboxypeptidase E/CPE affinity chromatography.
Catalog No. 50-161-1846