Learn More
Sino Biological ABCB5 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100692T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human ABCB5, and purified by antigen affinity chromatography.
Catalog No. 50-161-1820