Learn More
Sino Biological ACAN Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100693T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ACAN, and purified by antigen affinity chromatography.
Catalog No. 50-161-1824