Learn More
Sino Biological DDIT3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100710T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DDIT3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1862