Learn More
Sino Biological GNL3 / Nucleostemin Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100716T34200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GNL3 / Nucleostemin, and purified by antigen affinity chromatography.
Catalog No. 50-161-1871