Learn More
Sino Biological MSX1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100723T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MSX1, and purified by antigen affinity chromatography.
Catalog No. 50-161-1887