Learn More
Sino Biological Human CD14 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10073H08H50
A DNA sequence encoding the mature form of human CD14 (NP_000582.1) (Met 1-Met 344) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3945