Learn More
Sino Biological CD14 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10073MM01200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met 1-Met 344). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1919