Learn More
Sino Biological CD14 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10073RP01400
Produced in rabbits immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-1949