Learn More
Sino Biological CREB1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10074MM02200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CREB1 (rh CREB1; Catalog#10074-H10E; P16220-2; Met1-Asp327). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-1984