Learn More
Sino Biological CREB1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10074RP01400
Produced in rabbits immunized with purified, recombinant Human CREB1 (rh CREB1; Catalog#10074-H10E; P16220-2; Met1-Asp327). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-1991