Learn More
Sino Biological Annexin VI / ANXA6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100742T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human Annexin VI /ANXA6, and purified by antigen affinity chromatography.
Catalog No. 50-161-1961