Learn More
Sino Biological CAMKIV / CAMK4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100744T34200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CAMKIV/CAMK4 , and purified by antigen affinity chromatography.
Catalog No. 50-161-1966