Learn More
Sino Biological GOT2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100749T02200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GOT2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1981