Learn More
Sino Biological Human IL27 / Interleukin-27 Protein (His Tag), Biotinylated DFS Item
Manufacturer:  Sino Biological 10076H08SB10
A DNA sequence encoding the human IL27 complex composed of IL27B subunit (NP_005746.2) (Met1-Lys229) and the N-terminally truncated IL27p28 (IL27A) subunit (NP_663634.2) (Phe 29-Pro 243) linked by an polypeptide linker was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-161-3949