Learn More
Sino Biological IL-27 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10076MM02200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-27 (rh IL-27; Catalog#10076-H08S; NP_005746.2 & NP_663634.2; Met1-Lys229 & Phe29-Pro243). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-2038