Learn More
Sino Biological IL-27 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10076T16100
Produced in rabbits immunized with purified, recombinant Human IL-27 (rh IL-27; Catalog#10076-H08S; NP_005746.2 & NP_663634.2; Met1-Lys229 & Phe29-Pro243). IL-27 specific IgG was purified by Human IL-27 affinity chromatography.
Catalog No. 50-161-2045