Learn More
Sino Biological MBD4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100762T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MBD4, and purified by antigen affinity chromatography.
Catalog No. 50-161-2012