Learn More
Sino Biological Hdac6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100768T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Hdac6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2030