Learn More
Sino Biological CNDP1 / CN1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10077RP03100
Produced in rabbits immunized with purified, recombinant Human Carnosine Dipeptidase 1 (rhCNDP1; Catalog#10077-H08H; Ser 27-His 508; NP_116038.4). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-2077