Learn More
Sino Biological Aurora A / AURKA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100783T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Aurora A / AURKA, and purified by antigen affinity chromatography.
Catalog No. 50-161-2090