Learn More
Sino Biological Human Glypican 5 / GPC5 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10079H08B50
A DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3953