Learn More
Sino Biological HDAC4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100798T3850
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HDAC4, and purified by antigen affinity chromatography.
Catalog No. 50-161-2121