Learn More
Sino Biological PRMT1 / HRMT1L2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100805T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRMT1 / HRMT1L2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2137