Learn More
Sino Biological Human p38 alpha / MAPK14 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10081H07B20
A DNA sequence encoding the human MAPK14 isoform 2 (NP_620581.1) (Met1-Ser360) was fused with a polyhistide tag at the N-terminus.
Catalog No. 50-161-3954