Learn More
Sino Biological p38 alpha/MAPK14 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10081MM0750
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human p38 alpha/MAPK14 (rh p38 alpha/MAPK14; Catalog#10081-H07B; NP_620581.1; Met1-Ser360). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-2163