Learn More
Sino Biological p38 alpha / MAPK14 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10081T6050
Produced in rabbits immunized with purified, recombinant Human p38 alpha / MAPK14 (rh p38 alpha / MAPK14; Catalog#10081-H07B; NP_620581.1; Met1-Ser360). p38 alpha / MAPK14 specific IgG was purified by Human p38 alpha / MAPK14 affinity chromatography.
Catalog No. 50-161-2169