Learn More
Sino Biological Human MMP-2 / CLG4A Protein DFS Item
Manufacturer:  Sino Biological 10082HNAH50
A DNA sequence encoding the native human MMP2 (NP_004521.1) (Met 1-Cys 660) was expressed and purified.
Catalog No. 50-161-3959