Learn More
Sino Biological BCOR Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100827T0250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human BCOR, and purified by antigen affinity chromatography.
Catalog No. 50-161-2172