Learn More
Sino Biological GNAS Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100828T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GNAS, and purified by antigen affinity chromatography.
Catalog No. 50-161-2175