Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10084H02H200
A DNA sequence encoding the N-terminal segment (Met 1-Thr 239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-3961