Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein (ECD, Fc Tag) DFS Item
Manufacturer:  Sino Biological 10084H05H20
A DNA sequence encoding the human CD274 (NP_054862.1) (Met1-Thr239) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-3964