Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein (His Tag) , Biotinylated DFS Item
Manufacturer:  Sino Biological 10084H08HB50
A DNA sequence encoding the N-terminal segment (Met1-Thr239) of the extracellular domain of human B7-H1 (NP_054862.1) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-3967