Learn More
Sino Biological Human PD-L1 / B7-H1 / CD274 Protein DFS Item
Manufacturer:  Sino Biological 10084HNAH100
A DNA sequence encoding the human CD274 (NP_054862.1) (Met1-Arg238) was expressed.
Catalog No. 50-161-3968