Learn More
Sino Biological SMARCC1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100848T36200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SMARCC1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2220