Learn More
Sino Biological LRRTM2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100855T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LRRTM2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2285