Learn More
Sino Biological CD45 / PTPRC Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10086RP01100
Produced in rabbits immunized with purified, recombinant Human CD45 / PTPRC (rh PTPRC; Catalog#10086-H02H; NP_563578.1; Met1-Lys414). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-2313