Learn More
Sino Biological NCAPH Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100865T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NCAPH, and purified by antigen affinity chromatography.
Catalog No. 50-161-2292