Learn More
Sino Biological Human Glypican 3 / GPC3 / OCI-5 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10088H08H50
A DNA sequence encoding the human GPC3 (P51654-1) (Met 1-His 559) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-3973