Learn More
Sino Biological DAP3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100885T36200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DAP3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2329