Learn More
Sino Biological EGR1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100899T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human EGR1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2343