Learn More
Sino Biological ERCC8 / CSA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100906T34200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human ERCC8, and purified by antigen affinity chromatography.
Catalog No. 50-161-2359